siRNA Details
| Gene Symbol: LINC00675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036581.1 | 2 | 1097 | |||
| GenBank mRNA | BC015790.1 | - | 2 | 1035 | ||
| BC041634.1 | - | 2 | 1035 | |||
| BC063383.1 | - | 2 | 670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00675| Gene Symbol: LINC00675 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036581.1 | 2 | 1097 | |||
| GenBank mRNA | BC015790.1 | - | 2 | 1035 | ||
| BC041634.1 | - | 2 | 1035 | |||
| BC063383.1 | - | 2 | 670 | |||