siRNA Details
| Gene Symbol: SLC39A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252148.1 | NP_001239077.1 | 2, 3 | 914 | ||
| NM_001252150.1 | NP_001239079.1 | 2, 3 | 914 | |||
| NM_001252151.1 | NP_001239080.1 | 2, 3 | 341 | |||
| NM_018375.4 | NP_060845.2 | 2, 3 | 914 | |||
| GenBank mRNA | AK002136.1 | - | 2, 3 | 890 | ||
| AK222547.1 | - | 2, 3 | 892 | |||
| AK293222.1 | - | 2, 3 | 334 | |||
| AY358687.1 | - | 2, 3 | 360 | |||
| AY780789.1 | - | 2, 3 | 212 | |||
| BC047682.1 | - | 2, 3 | 844 | 6066186 | ||
| BC064383.1 | - | 2, 3 | 629 | 5737339 | ||
| BP269907.1 | - | 1, 2 | 116 | |||
| BX643103.1 | - | 2, 3 | 484 | |||
| FJ423549.1 | - | 2, 3 | 212 | |||

SLC39A9
Genomic Map