siRNA Details
Gene Symbol: LINC00683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120419.1 | 2 | 2103 | |||
GenBank mRNA | AK094957.1 | - | 2 | 1011 | ||
AK127565.1 | - | 2 | 2104 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120419.1 | 2 | 2103 | |||
GenBank mRNA | AK094957.1 | - | 2 | 1011 | ||
AK127565.1 | - | 2 | 2104 |