siRNA Details
| Gene Symbol: ZNF558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144693.2 | NP_653294.1 | 3 | 246 | ||
| XM_017026370.1 | XP_016881859.1 | 3 | 246 | |||
| XM_017026371.1 | XP_016881860.1 | 4 | 368 | |||
| XM_017026372.1 | XP_016881861.1 | 4 | 479 | |||
| GenBank mRNA | AK291265.1 | - | 3 | 201 | ||
| AK307200.1 | - | 3 | 241 | |||
| DB478966.1 | - | 3 | 376 | |||

ZNF558
Genomic Map