siRNA Details
| Gene Symbol: ZNF558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026371.1 | XP_016881860.1 | 2, 3 | 185 | ||
| XM_017026372.1 | XP_016881861.1 | 2, 3 | 296 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF558| Gene Symbol: ZNF558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026371.1 | XP_016881860.1 | 2, 3 | 185 | ||
| XM_017026372.1 | XP_016881861.1 | 2, 3 | 296 | |||