siRNA Details
| Gene Symbol: LINC00664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037194.1 | 8, 9 | 1578 | |||
| GenBank mRNA | AK097381.1 | - | 13, 14 | 1981 | ||
| BX537835.1 | - | 8, 9 | 1578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00664| Gene Symbol: LINC00664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037194.1 | 8, 9 | 1578 | |||
| GenBank mRNA | AK097381.1 | - | 13, 14 | 1981 | ||
| BX537835.1 | - | 8, 9 | 1578 | |||