siRNA Details
| Gene Symbol: LOC400706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125344.1 | 2 | 778 | |||
| GenBank mRNA | AK096622.1 | - | 2, 3 | 778 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC400706| Gene Symbol: LOC400706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125344.1 | 2 | 778 | |||
| GenBank mRNA | AK096622.1 | - | 2, 3 | 778 | ||