siRNA Details
| Gene Symbol: LOC400768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC051808.2 | - | 2 | 1886 | ||
| Gene Symbol: NBPF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011542012.2 | XP_011540314.1 | 2 | 1079 | ||

LOC400768
Genomic Map