siRNA Details
| Gene Symbol: LOC105371470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK097200.1 | - | 2 | 1124 | ||
| Gene Symbol: UTP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK055279.1 | - | 4 | 2078 | ||

LOC105371470
Genomic Map