siRNA Details
| Gene Symbol: LOC729296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109916.1 | 5 | 968 | |||
| NR_109917.1 | 5 | 738 | ||||
| NR_109918.1 | 5 | 635 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC729296| Gene Symbol: LOC729296 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109916.1 | 5 | 968 | |||
| NR_109917.1 | 5 | 738 | ||||
| NR_109918.1 | 5 | 635 | ||||