siRNA Details
| Gene Symbol: C22orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110522.1 | 3 | 1261 | |||
| NR_110523.1 | 5 | 1732 | ||||
| GenBank mRNA | BC033837.1 | - | 5 | 1705 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C22orf34| Gene Symbol: C22orf34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110522.1 | 3 | 1261 | |||
| NR_110523.1 | 5 | 1732 | ||||
| GenBank mRNA | BC033837.1 | - | 5 | 1705 | ||