siRNA Details
| Gene Symbol: LINC00954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022396.1 | - | 5 | 1237 | ||
| AK097927.1 | - | 6 | 1330 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00954| Gene Symbol: LINC00954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022396.1 | - | 5 | 1237 | ||
| AK097927.1 | - | 6 | 1330 | |||