siRNA Details
| Gene Symbol: LINC01159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110373.1 | 1 | 158 | |||
| NR_110374.1 | 1 | 158 | ||||
| GenBank mRNA | DB492871.1 | - | 1 | 161 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01159| Gene Symbol: LINC01159 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110373.1 | 1 | 158 | |||
| NR_110374.1 | 1 | 158 | ||||
| GenBank mRNA | DB492871.1 | - | 1 | 161 | ||