siRNA Details
| Gene Symbol: MBNL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027037.1 | 2 | 1078 | |||
| NR_027038.1 | 2 | 1089 | ||||
| GenBank mRNA | AK126417.1 | - | 1 | 175 | ||
| AK316515.1 | - | 2 | 1018 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MBNL1-AS1| Gene Symbol: MBNL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027037.1 | 2 | 1078 | |||
| NR_027038.1 | 2 | 1089 | ||||
| GenBank mRNA | AK126417.1 | - | 1 | 175 | ||
| AK316515.1 | - | 2 | 1018 | |||