siRNA Details
| Gene Symbol: LINC00884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033929.1 | 2 | 566 | |||
| GenBank mRNA | AK091527.1 | - | 2 | 553 | ||
| DA504887.1 | - | 2 | 553 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00884| Gene Symbol: LINC00884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033929.1 | 2 | 566 | |||
| GenBank mRNA | AK091527.1 | - | 2 | 553 | ||
| DA504887.1 | - | 2 | 553 | |||