siRNA Details
| Gene Symbol: LINC01511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125810.1 | 2 | 1525 | |||
| GenBank mRNA | BC034612.1 | - | 2 | 1517 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01511| Gene Symbol: LINC01511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125810.1 | 2 | 1525 | |||
| GenBank mRNA | BC034612.1 | - | 2 | 1517 | ||