siRNA Details
| Gene Symbol: PDK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002612.3 | NP_002603.1 | 6 | 963 | ||
| GenBank mRNA | AK096428.1 | - | 7 | 762 | ||
| AK098151.1 | - | 2 | 277 | |||
| AK223328.1 | - | 6 | 859 | |||
| AK312424.1 | - | 6 | 869 | |||
| BC040239.1 | - | 6 | 843 | 3047987 | ||
| U54617.1 | - | 6 | 865 | |||

PDK4
Genomic Map