siRNA Details
Gene Symbol: CARMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124798.1 | - | 1 | 459 | ||
AK126541.1 | - | 1 | 3566 | |||
BX640700.1 | - | 2 | 1845 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CARMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124798.1 | - | 1 | 459 | ||
AK126541.1 | - | 1 | 3566 | |||
BX640700.1 | - | 2 | 1845 |