siRNA Details
| Gene Symbol: STMND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190766.1 | NP_001177695.1 | 4, 5 | 539 | ||
| GenBank mRNA | AK026805.1 | - | 1, 2 | 1302 | ||
| DT932725.1 | - | 3, 4 | 379 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

STMND1| Gene Symbol: STMND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190766.1 | NP_001177695.1 | 4, 5 | 539 | ||
| GenBank mRNA | AK026805.1 | - | 1, 2 | 1302 | ||
| DT932725.1 | - | 3, 4 | 379 | |||