siRNA Details
| Gene Symbol: CASC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015410.1 | 5 | 740 | |||
| GenBank mRNA | AK022865.1 | - | 4 | 366 | ||
| AK026189.1 | - | 5 | 740 | |||
| AK096633.1 | - | 3 | 339 | |||
| AK300802.1 | - | 6 | 895 | |||
| AK310212.1 | - | 7 | 1044 | |||
| BC044235.1 | - | 2 | 189 | |||
| KP981381.1 | - | 2 | 254 | |||

CASC15
Genomic Map