siRNA Details
| Gene Symbol: CASC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015410.1 | 3, 4 | 540 | |||
| GenBank mRNA | AK022865.1 | - | 2, 3 | 166 | ||
| AK026189.1 | - | 3, 4 | 540 | |||
| AK300802.1 | - | 4, 5 | 695 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASC15| Gene Symbol: CASC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015410.1 | 3, 4 | 540 | |||
| GenBank mRNA | AK022865.1 | - | 2, 3 | 166 | ||
| AK026189.1 | - | 3, 4 | 540 | |||
| AK300802.1 | - | 4, 5 | 695 | |||