siRNA Details
| Gene Symbol: TRAM2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103446.1 | 1 | 781 | |||
| GenBank mRNA | AK095117.1 | - | 1 | 676 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRAM2-AS1| Gene Symbol: TRAM2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103446.1 | 1 | 781 | |||
| GenBank mRNA | AK095117.1 | - | 1 | 676 | ||