siRNA Details
| Gene Symbol: TSTD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195131.1 | NP_001182060.1 | 4 | 1190 | ||
| GenBank mRNA | BX647075.1 | - | 5 | 1276 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSTD3| Gene Symbol: TSTD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195131.1 | NP_001182060.1 | 4 | 1190 | ||
| GenBank mRNA | BX647075.1 | - | 5 | 1276 | ||