siRNA Details
| Gene Symbol: ZNF815P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023382.1 | 5 | 1308 | |||
| GenBank mRNA | AK096288.1 | - | 5 | 1307 | ||
| Gene Symbol: ZNF890P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034163.1 | 6 | 1299 | |||
| GenBank mRNA | AK295403.1 | - | 6 | 1299 | ||

ZNF815P
Genomic Map