siRNA Details
| Gene Symbol: LINC00998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015442.1 | 3 | 714 | |||
| NR_024412.1 | 3 | 802 | ||||
| GenBank mRNA | BC065765.1 | - | 3 | 690 | ||
| BC107860.1 | - | 3 | 718 | |||
| BX537645.1 | - | 3 | 687 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00998| Gene Symbol: LINC00998 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015442.1 | 3 | 714 | |||
| NR_024412.1 | 3 | 802 | ||||
| GenBank mRNA | BC065765.1 | - | 3 | 690 | ||
| BC107860.1 | - | 3 | 718 | |||
| BX537645.1 | - | 3 | 687 | |||