siRNA Details
| Gene Symbol: LOC107984125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014157.1 | XP_016869646.1 | 20 | 2835 | ||
| XM_017014158.1 | XP_016869647.1 | 15 | 2171 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107984125| Gene Symbol: LOC107984125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017014157.1 | XP_016869646.1 | 20 | 2835 | ||
| XM_017014158.1 | XP_016869647.1 | 15 | 2171 | |||