siRNA Details
| Gene Symbol: LINC00967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039979.1 | 3 | 1156 | |||
| GenBank mRNA | BX389854.2 | - | 2 | 302 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00967| Gene Symbol: LINC00967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039979.1 | 3 | 1156 | |||
| GenBank mRNA | BX389854.2 | - | 2 | 302 | ||