siRNA Details
| Gene Symbol: LINC01285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110393.1 | 6 | 577 | |||
| GenBank mRNA | AK057519.1 | - | 6 | 577 | ||
| DB165975.1 | - | 6 | 541 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01285| Gene Symbol: LINC01285 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110393.1 | 6 | 577 | |||
| GenBank mRNA | AK057519.1 | - | 6 | 577 | ||
| DB165975.1 | - | 6 | 541 | |||