siRNA Details
| Gene Symbol: ZC3H15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018471.2 | NP_060941.2 | 2 | 339 | ||
| XM_017004502.1 | XP_016859991.1 | 2 | 244 | |||
| GenBank mRNA | AF109366.1 | - | 2 | 198 | ||
| AF161421.1 | - | 2 | 210 | |||
| AF220184.1 | - | 2 | 340 | |||
| AK001901.1 | - | 2 | 244 | |||
| AK297658.1 | - | 2 | 244 | |||
| AK298615.1 | - | 293 | ||||
| BC021102.1 | - | 2 | 151 | 5020890 | ||

ZC3H15
Genomic Map