siRNA Details
| Gene Symbol: SLC22A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001307985.1 | NP_001294914.1 | 5 | 1265 | ||
| NM_018484.3 | NP_060954.1 | 5 | 1265 | |||
| XM_011545167.1 | XP_011543469.1 | 4 | 915 | |||
| GenBank mRNA | AB026116.1 | - | 5 | 918 | ||
| AK075127.1 | - | 3 | 291 | |||
| AK075224.1 | - | 5 | 611 | |||
| AK129930.1 | - | 5 | 1177 | |||
| AK222869.1 | - | 5 | 923 | |||
| AK290791.1 | - | 5 | 1248 | |||
| AK309931.1 | - | 5 | 925 | |||
| BC034384.1 | - | 5 | 1240 | 5190509 | ||

SLC22A11
Genomic Map