siRNA Details
| Gene Symbol: FLJ32742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057304.1 | - | 2 | 1523 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ32742| Gene Symbol: FLJ32742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK057304.1 | - | 2 | 1523 | ||