siRNA Details
| Gene Symbol: SLC22A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033396.1 | 10 | 1707 | |||
| XM_017017775.1 | XP_016873264.1 | 7 | 1514 | |||
| GenBank mRNA | AK094565.1 | - | 4 | 1226 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A20| Gene Symbol: SLC22A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033396.1 | 10 | 1707 | |||
| XM_017017775.1 | XP_016873264.1 | 7 | 1514 | |||
| GenBank mRNA | AK094565.1 | - | 4 | 1226 | ||