siRNA Details
Gene Symbol: SLC22A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033396.1 | 10 | 2218 | |||
XM_017017775.1 | XP_016873264.1 | 7 | 2025 | |||
GenBank mRNA | AK094565.1 | - | 4 | 1737 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLC22A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033396.1 | 10 | 2218 | |||
XM_017017775.1 | XP_016873264.1 | 7 | 2025 | |||
GenBank mRNA | AK094565.1 | - | 4 | 1737 |