siRNA Details
| Gene Symbol: SLC22A20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033396.1 | 7 | 1165 | |||
| XM_017017774.1 | XP_016873263.1 | 6 | 996 | |||
| XM_017017775.1 | XP_016873264.1 | 4 | 972 | |||
| XM_017017776.1 | XP_016873265.1 | 4 | 972 | |||
| GenBank mRNA | DN831899.1 | - | 3 | 260 | ||

SLC22A20
Genomic Map