siRNA Details
| Gene Symbol: LRCOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195520.1 | NP_001182449.1 | 3 | 296 | ||
| XM_011534780.2 | XP_011533082.1 | 3 | 296 | |||
| XM_017018659.1 | XP_016874148.1 | 2 | 272 | |||
| XM_017018660.1 | XP_016874149.1 | 2 | 272 | |||
| GenBank mRNA | BX282338.1 | - | 3 | 242 | ||
| JQ012741.1 | - | 3 | 219 | |||
| JQ012742.1 | - | 3 | 351 | |||

LRCOL1
Genomic Map