siRNA Details
| Gene Symbol: LRCOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534780.2 | XP_011533082.1 | 5 | 678 | ||
| XM_011534784.2 | XP_011533086.1 | 4 | 536 | |||
| XM_011534785.2 | XP_011533087.1 | 4 | 541 | |||
| XM_017018659.1 | XP_016874148.1 | 4 | 654 | |||
| XM_017018661.1 | XP_016874150.1 | 4 | 896 | |||
| XM_017018662.1 | XP_016874151.1 | 4 | 639 | |||

LRCOL1
Genomic Map