siRNA Details
| Gene Symbol: ZNF891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277291.1 | NP_001264220.1 | 2 | 2035 | ||
| XM_017018666.1 | XP_016874155.1 | 2 | 2203 | |||
| XM_017018667.1 | XP_016874156.1 | 2 | 1873 | |||
| XM_017018668.1 | XP_016874157.1 | 2 | 1956 | |||

ZNF891
Genomic Map