siRNA Details
| Gene Symbol: SMIM10L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271592.1 | NP_001258521.1 | 1 | 684 | ||
| GenBank mRNA | BG680492.1 | - | 1 | 624 | ||
| BX338800.2 | - | 1 | 650 | |||
| CX164259.1 | - | 1 | 618 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMIM10L1| Gene Symbol: SMIM10L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271592.1 | NP_001258521.1 | 1 | 684 | ||
| GenBank mRNA | BG680492.1 | - | 1 | 624 | ||
| BX338800.2 | - | 1 | 650 | |||
| CX164259.1 | - | 1 | 618 | |||