siRNA Details
| Gene Symbol: IL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018402.1 | NP_060872.1 | 5 | 485 | ||
| GenBank mRNA | AJ251549.1 | - | 5 | 485 | ||
| BC066270.1 | - | 5 | 484 | 7002121 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL26| Gene Symbol: IL26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018402.1 | NP_060872.1 | 5 | 485 | ||
| GenBank mRNA | AJ251549.1 | - | 5 | 485 | ||
| BC066270.1 | - | 5 | 484 | 7002121 | ||