siRNA Details
| Gene Symbol: GOLGA8M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282468.1 | NP_001269397.1 | 19 | 2844 | ||
| XM_005268284.4 | XP_005268341.1 | 19 | 3643 | |||
| XM_005268285.4 | XP_005268342.1 | 18 | 3574 | |||
| XM_006720654.3 | XP_006720717.1 | 19 | 2787 | |||
| XM_011521951.2 | XP_011520253.1 | 20 | 3212 | |||
| XM_011521954.2 | XP_011520256.1 | 19 | 2777 | |||
| Gene Symbol: GOLGA8K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282493.1 | NP_001269422.1 | 19 | 2838 | ||
| Gene Symbol: GOLGA8N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282494.1 | NP_001269423.1 | 19 | 2844 | ||
| Gene Symbol: GOLGA8O | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277308.1 | NP_001264237.1 | 19 | 2844 | ||
| XM_011521988.2 | XP_011520290.1 | 18 | 3460 | |||
| Gene Symbol: GOLGA8R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282484.1 | NP_001269413.1 | 19 | 2841 | ||
| Gene Symbol: GOLGA8S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038843.1 | 18 | 3111 | |||
| Gene Symbol: GOLGA8T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033933.1 | 17 | 2693 | |||

GOLGA8M
Genomic Map