siRNA Details
| Gene Symbol: LOC100288637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038253.1 | 6 | 756 | |||
| NR_038254.1 | 5 | 656 | ||||
| NR_038255.1 | 4 | 561 | ||||
| GenBank mRNA | BC112921.1 | - | 4 | 433 | ||
| BX954662.1 | - | 4 | 539 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100288637| Gene Symbol: LOC100288637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038253.1 | 6 | 756 | |||
| NR_038254.1 | 5 | 656 | ||||
| NR_038255.1 | 4 | 561 | ||||
| GenBank mRNA | BC112921.1 | - | 4 | 433 | ||
| BX954662.1 | - | 4 | 539 | |||