siRNA Details
| Gene Symbol: ZSCAN29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006720401.3 | XP_006720464.1 | 2 | 560 | ||
| XM_017021949.1 | XP_016877438.1 | 1 | 772 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZSCAN29| Gene Symbol: ZSCAN29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006720401.3 | XP_006720464.1 | 2 | 560 | ||
| XM_017021949.1 | XP_016877438.1 | 1 | 772 | |||