siRNA Details
Gene Symbol: LOC145783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015419.2 | 1 | 864 | |||
GenBank mRNA | AK056606.1 | - | 1 | 13 | ||
AK096657.1 | - | 1 | 865 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC145783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015419.2 | 1 | 864 | |||
GenBank mRNA | AK056606.1 | - | 1 | 13 | ||
AK096657.1 | - | 1 | 865 |