siRNA Details
| Gene Symbol: LOC102723493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135686.1 | 4 | 1661 | |||
| GenBank mRNA | BX106667.1 | - | 3 | 577 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102723493| Gene Symbol: LOC102723493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135686.1 | 4 | 1661 | |||
| GenBank mRNA | BX106667.1 | - | 3 | 577 | ||