siRNA Details
Gene Symbol: LOC102723493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135686.1 | 3 | 1553 | |||
GenBank mRNA | BX106667.1 | - | 2 | 469 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102723493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135686.1 | 3 | 1553 | |||
GenBank mRNA | BX106667.1 | - | 2 | 469 |