siRNA Details
Gene Symbol: DNM1P41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033787.2 | 1 | 278 |
Gene Symbol: LOC102724034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120378.1 | 3 | 519 | |||
GenBank mRNA | DC422276.1 | - | 3 | 519 |
Gene Symbol: LOC103171574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120379.1 | 3 | 618 | |||
GenBank mRNA | AK311032.1 | - | 3 | 519 |
