siRNA Details
| Gene Symbol: LOC102724034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120378.1 | 3 | 485 | |||
| GenBank mRNA | DC422276.1 | - | 3 | 485 | ||
| Gene Symbol: LOC103171574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120379.1 | 3 | 584 | |||
| GenBank mRNA | AK311032.1 | - | 3 | 485 | ||

LOC102724034
Genomic Map