siRNA Details
| Gene Symbol: LOC101927751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135827.1 | 1 | 410 | |||
| NR_135828.1 | 1 | 410 | ||||
| NR_135831.1 | 1 | 410 | ||||
| GenBank mRNA | AK126286.1 | - | 1 | 410 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927751| Gene Symbol: LOC101927751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135827.1 | 1 | 410 | |||
| NR_135828.1 | 1 | 410 | ||||
| NR_135831.1 | 1 | 410 | ||||
| GenBank mRNA | AK126286.1 | - | 1 | 410 | ||