siRNA Details
| Gene Symbol: LOC100506990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK129982.1 | - | 5 | 1317 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506990| Gene Symbol: LOC100506990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK129982.1 | - | 5 | 1317 | ||