siRNA Details
| Gene Symbol: FBRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105079.2 | NP_001098549.2 | 3, 4 | 672 | ||
| XM_011545916.2 | XP_011544218.1 | 3, 4 | 2049 | |||
| GenBank mRNA | DB082271.1 | - | 3, 4 | 269 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBRS| Gene Symbol: FBRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105079.2 | NP_001098549.2 | 3, 4 | 672 | ||
| XM_011545916.2 | XP_011544218.1 | 3, 4 | 2049 | |||
| GenBank mRNA | DB082271.1 | - | 3, 4 | 269 | ||